Få mere at vide
EN 14039/ISO 16703 Mineral Oil Mixture 234 4000 μg/mL in Heptane, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diesel fuel No.2∣Mineral Oil (without additives) |
| CAS | 68476-34-6, 8042-47-5 |
| Analyt- eller komponentnavne | Diesel fuel No.2∣Mineral Oil (without additives) |
| Forsendelsestilstand | Room Temperature |
| Certificeringer/compliance | ISO 17034 |
| Opløsningsmiddel eller Matrix | Heptane |
| Industry Type | Food Testing |
| Mængde | 5 x 1 ml |
| Koncentration eller sammensætning (efter analyt eller komponenter) | Diesel fuel No.2 4000 μg/mL∣Mineral Oil (without additives) 4000 μg/mL |