Få mere at vide
Emylcamate, LoGiCal
Artikelnummer. 30464045
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464045 10 mg
1 options
Artikelnummer. 30464045 Leverandør LoGiCal Leverandørnr. LGCFOR3364.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Emylcamate
CAS 78-28-4
Analyt- eller komponentnavne Emylcamate
Forsendelsestilstand Room Temperature
Molekylær formel C7 H15 N O2
Certificeringer/compliance ISO 9001
Synonym 3-Pentanol, 3-methyl-, 3-carbamate, 3-Pentanol, 3-methyl-, carbamate (8CI,9CI), Carbamic acid, 1-ethyl-1-methylpropyl ester (6CI,7CI), 1-Ethyl-1-methylpropyl carbamate, 3-Methyl-3-pentanol carbamate, 91 JD, Diethyl methyl carbinol urethan, Emylcamate, JD 91, KABI 925, MK 250, Methyl diethyl carbinol urethan, NSC 169882, NSC 169917, Nuncital, Restetal, Statran, Striatran, tert-Hexanol carbamate
InChI formel InChI=1S/C7H15NO2/c1-4-7(3,5-2)10-6(8)9/h4-5H2,1-3H3,(H2,8,9)
Løsningstype Neat
SMIL CCC(C)(CC)OC(=O)N
IUPAC navn 3-methylpentan-3-yl carbamate
Industry Type Clinical Testing
Molekylvægt (g/mol) 145.2
Mængde 10 mg
Formel vægt 145.1103 g/mol