missing translation for 'onlineSavingsMsg'
Få mere at vide

EMPA, TRC

Artikelnummer. 30380391
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30380391 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380391 Supplier Toronto Research Chemicals Supplier No. TRCE52155010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale EMPA
CAS 680590-49-2
Molekylær formel C23H26N4O4S
Mængde 10 mg
Synonym N-Ethyl-2-[(6-methoxy-3-pyridinyl)[(2-methylphenyl)sulfonyl]amino]-N-(3-pyridinylmethyl)-acetamide
InChI formel InChI=1S/C23H26N4O4S/c1-4-26(16-19-9-7-13-24-14-19)23(28)17-27(20-11-12-22(31-3)25-15-20)32(29,30)21-10-6-5-8-18(21)2/h5-15H,4,16-17H2,1-3H3
SMIL O=C(N(CC=1C=NC=CC1)CC)CN(C2=CN=C(OC)C=C2)S(=O)(=O)C=3C=CC=CC3C
IUPAC navn N-ethyl-2-[(6-methoxypyridin-3-yl)-(2-methylphenyl)sulfonylamino]-N-(pyridin-3-ylmethyl)acetamide
Molekylvægt (g/mol) 454.54
Formel vægt 454.1675
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.