Få mere at vide
Emodin-d4, TRC
Artikelnummer. 30368877
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30368877 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368877 Leverandør Toronto Research Chemicals Leverandørnr. TRCE52300210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Emodin-d4
CAS 132796-52-2
Molekylær formel C15H6D4O5
Mængde 10 mg
Synonym 1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione-d4, 1,6,8-Trihydroxy-3-methylanthraquinone-d4, 4,5,7-Trihydroxy-2-methyl-
InChI formel InChI=1S/C15H10O5/c1-6-2-8-12(10(17)3-6)15(20)13-9(14(8)19)4-7(16)5-11(13)18/h2-5,16-18H,1H3/i2D,3D,4D,5D
SMIL OC1=C([2H])C(C)=C([2H])C(C(C2=C3C(O)=C([2H])C(O)=C2[2H])=O)=C1C3=O
Molekylvægt (g/mol) 274.26
Formel vægt 274.08