Få mere at vide
EML1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
4670.00 DKK
Tekniske data
| Antigen | EML1 |
|---|---|
| Fortynding | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applikationer | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
Beskrivelse
EML1 Polyclonal specifically detects EML1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Tekniske data
| EML1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| echinoderm microtubule associated protein like 1, ELP79, EMAP1, EMAP-1, EMAPechinoderm microtubule-associated protein-like 1, EMAPL1, EMAPLhuEMAP-1, FLJ45033, HuEMAP, HuEMAP-1 | |
| EML1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O00423 | |
| 2009 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SGVRKETAVPATKSNIKRTSSSERVSPGGRRESNGDSRGNRNRTGSTSSS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |