Få mere at vide
Emetine Hydrochloride Hydrate, TRC
Artikelnummer. 30378754
Skift visning
Mængde:
10 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378754 250 mg
30375118 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378754 Leverandør Toronto Research Chemicals Leverandørnr. TRCE521535250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7083-71-8
Molekylær formel C29 H40 N2 O4 . 2 Cl H . H2 O
Molekylvægt (g/mol) 571.58
Synonym Emetan, 6',7',10,11-tetramethoxy-, dihydrochloride, hydrate, Emetine, dihydrochloride, hydrate (8CI), 2H-Benzo[a]quinolizine, emetan deriv., Emetine hydrochloride hydrate
IUPAC navn (2S,3R,11bS)-2-[[(1R)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl]methyl]-3-ethyl-9,10-dimethoxy-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizine;hydrate;dihydrochloride
SMIL O.Cl.Cl.CC[C@H]1CN2CCc3cc(OC)c(OC)cc3[C@@H]2C[C@@H]1C[C@H]4NCCc5cc(OC)c(OC)cc45
Tekniske data
Kemisk navn eller materiale Emetine dihydrochloride hydrate
Mængde 250 mg
InChI formel InChI=1S/C29H40N2O4.2ClH.H2O/c1-6-18-17-31-10-8-20-14-27(33-3)29(35-5)16-23(20)25(31)12-21(18)11-24-22-15-28(34-4)26(32-2)13-19(22)7-9-30-24;;;/h13-16,18,21,24-25,30H,6-12,17H2,1-5H3;2*1H;1H2/t18-,21-,24+,25-;;;/m0.../s1
Formel vægt 570.2627