Learn More
Merck Accessories for Sterifil™ Aseptic Systems
Used for general filtration applications and filtration of samples for particulate or biological contamination analysis
109.00 DKK - 764.00 DKK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10019810
|
Merck
XX1104702 |
Base Holder with Support Screen |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10529351
|
Merck
XX1104705 |
250 mL Receiver Flask |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10603212
|
Merck
XX1104703 |
Funnel Cover |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10263251
|
Merck
XX1104706 |
Flask Cover |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
MilliporeSigma Polypropylene Support Screen, For Use With: For Swinnex 47, Surface Area: Support Screen, 47mm dia. Filter, Polypropylene, for Swinnex 47
For use with Sterifil™ Aseptic SystemsOrder Info
The Sterifil holder and funnel available separately (without receiver flask and cover) to use with standard 1L vacuum filtering flask or multiple place manifold.
Specifications
Not Autoclavable |