Learn More
Merck Millipore Stainless-Steel Pressure Filter Holders
Ultraclean or sterilize liquids or gases by pressure filtration
11312.00 DKK - 18585.00 DKK
Specifications
Description Filter Holder
View More Specs
Products 2
Product Code Brand Diameter (English) Inside Height (English) Material Price Quantity & Availability  
Product Code Brand Diameter (English) Inside Height (English) Material Price Quantity & Availability  
10430031
View accessory products View Documents
Merck Millipore
YY3009000
4.75 in. 6.75 in. PTFE
11312.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment
03-05-2021
Add to basket
10054510
View accessory products View Documents
Merck Millipore
YY3014236
7.25 in. 10.62 in. Stainless Steel
18585.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment
30-04-2021
Add to basket
Description
Description
  • Autoclavable with MF (mixed cellulose esters) filter in place
  • Type 316 stainless steel
  • Anodized aluminum legs
  • PTFE O-rings
90mm Filter
  • 1/4 NPTF inlet
  • Outlet with connections supplied for 0.87 in. I.D. (9.5mm) hose
  • 1/8 NPTF upstream port for vent/relief valve
142mm Filter
  • 11/8 NPTF TC flange with clamps and adapters supplied for 0.56 in. I.D. (14mm) hose
  • 1/8 NPTF upstream port for vent/relief
Other O-ring materials are available, as well as accessory cylinders to convert holders into pressure cells without separate reservoirs. Call your Fisher Scientific™ Customer Service Representative for details.
Specifications
Specifications
Filter Holder
SDS