Description
- Sterile, polystyrene
- Can be used as receivers for Millipore Steritop™ Vacuum Bottle-Top Filters
- 45mm neck with screw-top lid
- Storage of sterilized or clarified fluids
- Laboratory Filtration
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Merck™ Replacement Receiver Flasks