Learn More
Merck MultiScreen™ Multiple Punch Kits
Includes punch with punch carrier plate and punch distributor
Brand:  Merck MAPK8960A
41101.75 DKK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11556365
  • 45455.00 DKK / Each
Estimated Shipment: 11-07-2024
to see stock.
Specifications
Radionucleotide counting by transfer of individual filters to tubes
Multiple punch, 2 carrier racks for 7mL vials
1/Pk.