Learn More
Merck Millipore Milli-Q™ Series Water Purification System Accessory, Millipak™ 40 Express Final Filter
Use for the production of bacteria-free purified water
Brand:  Merck Millipore MPGP04001
Additional Details : Weight : 0.11700kg
Product Code. 12809355
  • 3240.00 DKK / Each
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
Use for the production of bacteria-free purified water
  • Pore size: 0.22μm
  • Non-sterile
    • Certifications
      ISO 9001 v.2000 and ISO 14001
      Compliance
      USP 645 suitability requirements