Learn More
Merck Microscope Slides
Product Code. 10161681
Change view
Click to view available options
2 x 3 in. dimensions
Product Code.
10161681 2 x 3 in. dimensions
1 options
Product Code. 10161681 Supplier Merck Supplier No. XX1007615
  • ONLINE EXCLUSIVE PRICE
    958.00 DKK
    744.52 DKK / Pakke med 72
    Spare 213.48 DKK
    22% Slukket
Forventet forsendelse: 18-12-2026
Accessories for filter holders for sample preparation.
Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY
Specifications
Width (Metric) 5.0 cm
No. per Pack 72
Sterility Non-sterile
Frosted Not Frosted
Material Glass
Type Microscope Slide
Well Count 0
Color Clear
Coating Uncoated
For Use With (Application) Accessories for filter holders for sample preparation
Length (English) 2 in.
Width (English) 2 in.
Dimensions (L x W) 2 x 3 in.