Få mere at vide
Merck Millipore Microfil™ V Filtration Device
Filter water and other beverages. Microfil™ V Filtration Device is a sterile filter that can contain 100mL.
Indeholder: Filtration device; 100mL polypropylene funnel; white, gridded sterile 47mm MCE membrane with 0.22 or 0.45μm pore size.
Beskrivelse
Uniform microorganism recovery is promoted by unique “push-fit” seal; smooth, hydrophobic funnel surface reduces aqueous sample residues sticking to funnel walls; and support's convex shape eliminates membrane distortion, thus maintaining uniform contact between membrane and medium surface
- No need to assemble filter holders, clean funnels, or sterilize filter support after each filtration
- Microfil V filtration device snaps off the carrying tray and onto a standard No. 8 stopper or a vacuum syringe with Luer connection
- Funnels attach with “push-fit” seal to prevent sample bypass without clamps or O-rings
- Filter support has a curved edge to easily remove membrane after filtration while vacuum is still applied
Ordreinfo
Sold in three trays of eight
Certificeringer
Membrane is certified in accordance with Standard Methods for Coliform Analysis.
Overholdelse
Conforms to international standards for testing drinking water, Standard Methods for the Examination of Water and Wastewater, and WHO Guidelines for Drinking Water Quality.