missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Microbiological Analysis Membrane Filters

Optimized for MF method microbiological analysis of water or other liquids

Brand:  Merck Millipore HAWP047S0

Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 13187654

Quantity Price
1 1788.00 DKK / Pack of 100
6 1519.80 DKK / Pack of 100
Estimated Shipment
21-05-2021
Add to basket


Description

Description

  • MF-Type (mixed cellulose esters) filter membranes
  • 0.45μm (Type HA) designed to give complete retention and maximum recovery of total coliforms and fecal coliform bacteria
  • Use 0.22μm membranes where 0.45μm pore size is not sufficient
  • For beverage analysis, choose membranes with 0.8μm pore size

Compliance

Certified to meet Standard Methods for the Examination of Water and Wastewater requirements.

Specifications

Specifications

Certified to meet standard methods for the examination of Water and wastewater requirements
1.85 in.
MF method microbiological analysis of water or other liquids
Plain
0.45 μm
White
4.7 cm
Absorbent pads
SO Pak Membrane Filter
Sterile
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 0.45µm 25mm white plain S0-pak filter membrane made of mixed ester cellulose

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose, octylphenol polyethoxyethanol

SDS
Documents