Learn More
Merck Millipore Filter Holder Vacuum Manifolds
Provide vacuum support for simultaneous filtration of either three or six test samples
Brand:  Merck Millipore XX2504700
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 11522124
  • POA / Each
Currently not in stock
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on +45 70 27 99 20 for assistance.
Description
Description
  • Use Millipore glass, plastic or stainless-steel filter holders fitted with standard No. 8 perforated stoppers over the holder outlets
  • Manifolds connect to single vacuum source and use separate control valves at each station for independent operation
  • Use vent ports on each stainless-steel manifold station to attach Swinnex™ filter holders or Millex™ filter units to maintain sterility on the test filter's downstream side
Stainless-steel Manifolds
  • Stainless-steel body, three-way valves, filter holder supports; anodized aluminum handles; stainless-steel hose connector, pipe plug
  • Units are autoclavable at 121°C (250°F)
PVC Manifold
  • Polyvinylchloride body; stainless-steel two-way valve; polypropylene hose adapters and handles
  • Not autoclavable
Specifications
Specifications
Manifold
Vacuum Filter Holder
6
Stainless Steel