missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore EZ-Pak™ Membranes

Used for general microbiological analysis. EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.

Brand:  Merck Millipore EZAAWG474

Additional Details : Weight : 0.70000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 13157174

  • 4275.00 DKK / Pack of 600


Estimated Shipment
07-06-2021
Add to basket

Description

Description

Used for general microbiological analysis. EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.

Press lever to uncover half of membrane surface for easy retrieval with forceps; other half remains in protective packaging. Unused membranes remain safely packed in pleated band.

  • Mixed Cellulose Ester
  • Sterile and grid surface
  • Used for general microbiological analysis
  • Certified to meet standard methods for the examination of Water and wastewater requirements

Specifications

Specifications

Certified to meet standard methods for the examination of Water and wastewater requirements
1.85 in.
Particle counting, yeast or mold
Grid
Mixed Cellulose Ester
Sterile
White
4.7 cm
Forceps
EZ-Pak Membrane
0.8 μm
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 0.8µm 47mm white gridded EZ-pak filter membrane made of mixed ester cellulose

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose, octylphenol polyethoxyethanol

SDS
Documents