Learn More
Merck Millipore Elastase, Human Neutrophil, Protein
Brand:  Merck Millipore 324681-100UG
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12888355
  • 4852.00 DKK / 100µg
Estimated Shipment
18-03-2021
Description
Description
Native elastase from human neutrophil. A serine protease that degrades elastin, collagen, and proteoglycans. Its overexpression has been implicated in the development of pulmonary emphysema, rheumatoid arthritis, and cystic fibrosis.
Specifications
Specifications
Lyophilized Solid
Reconstitute in 50mM sodium acetate, pH 5.5, with 200mM NaCl or in other aqueous buffers. Material is most active near a neutral pH, but to maintain stability upon reconstitution, use in a slightly acidic buffer (pH 5.5) to avoid autolysis.
100μg
Following reconstitution, aliquot and freeze (−20°C) for long term storage or refrigerate (4°C) for short-term storage. Acidic stock solutions are stable for up to 1 month at 4°C or for up to 1 year at −20°C.
Human Neutrophil
Salt-free.
Major band by SDS-PAGE
Elastase
RUO
No