Få mere at vide
Tocris Bioscience™ EMD 281014 hydrochloride
Artikelnummer. 30001585
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001585 50 mg
30001548 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001585 Leverandør Tocris Bioscience™ Leverandørnr. 4470/50
Selective 5-HT2A antagonist
Selective 5-HT2A antagonist; displays high affinity for human 5-HT2A over 5-HT2C receptors (Ki values are 0.87 and 557 nM respectively). Exhibits low off-target activity (Ki values are >1000 and 1268 nM for human D2 receptors and IKr channels, respectively).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 443144-27-2
Molekylær formel C22H22ClFN4O
Tekniske data
Kemisk navn eller materiale EMD 281014 hydrochloride
Mængde 50 mg
Formel vægt 412.89
Renhed >98%
Molekylvægt (g/mol) 412.893