Få mere at vide
Embutramide, LoGiCal
Artikelnummer. 30463730
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463730 10 mg
1 options
Artikelnummer. 30463730 Leverandør LoGiCal Leverandørnr. LGCFOR3065.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Embutramide
CAS 15687-14-6
Analyt- eller komponentnavne Embutramide
Forsendelsestilstand Room Temperature
Molekylær formel C17 H27 N O3
Certificeringer/compliance ISO 9001
Synonym Butanamide, N-[2-ethyl-2-(3-methoxyphenyl)butyl]-4-hydroxy-, Embutane, Embutramide, HOE 18 680, HOE 18680, T 61, Butyramide, N-(beta,beta-diethyl-m-methoxyphenethyl)-4-hydroxy- (8CI), N-[2-Ethyl-2-(3-methoxyphenyl)butyl]-4-hydroxybutanamide, N-(beta,beta-Diethyl-m-methoxyphenethyl)-4-hydroxybutyramide, N-[2-(m-Methoxyphenyl)-2-ethylbutyl]-gamma-hydroxybutyramide
InChI formel InChI=1S/C17H27NO3/c1-4-17(5-2,13-18-16(20)10-7-11-19)14-8-6-9-15(12-14)21-3/h6,8-9,12,19H,4-5,7,10-11,13H2,1-3H3,(H,18,20)
Løsningstype Neat
SMIL CCC(CC)(CNC(=O)CCCO)c1cccc(OC)c1
IUPAC navn N-[2-ethyl-2-(3-methoxyphenyl)butyl]-4-hydroxybutanamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 293.4
Mængde 10 mg
Formel vægt 293.1991 g/mol