missing translation for 'onlineSavingsMsg'
Få mere at vide

Embramine, TRC

Artikelnummer. 30372374
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30372374 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372374 Supplier Toronto Research Chemicals Supplier No. TRCE521005250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Embramine
CAS 3565-72-8
Molekylær formel C18H22BrNO
Mængde 250 mg
Synonym 2-[1-(4-Bromophenyl)-1-phenylethoxy]-N,N-dimethylethanamine, 1-(2-Dimethylaminoethoxy)-1-(p-bromophenyl)-1-phenylethane, 2-[(p-Bromo-alpha-methyl-alpha-phenylbenzyl)oxy]-N,N-dimethyl Ethylamine
InChI formel InChI=1S/C18H22BrNO/c1-18(21-14-13-20(2)3,15-7-5-4-6-8-15)16-9-11-17(19)12-10-16/h4-12H,13-14H2,1-3H3
SMIL CN(C)CCOC(C)(c1ccccc1)c2ccc(Br)cc2
IUPAC navn 2-[1-(4-bromophenyl)-1-phenylethoxy]-N,N-dimethylethanamine
Molekylvægt (g/mol) 348.28
Formel vægt 347.0885
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.