Få mere at vide
Embramine Hydrochloride, LoGiCal
Artikelnummer. 30460965
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30460965 5 mg
1 options
Artikelnummer. 30460965 Leverandør LoGiCal Leverandørnr. LGCFOR1971.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Embramine Hydrochloride
CAS 13977-28-1
Analyt- eller komponentnavne Embramine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C18 H22 Br N O . Cl H
Certificeringer/compliance ISO 9001
Synonym Ethanamine, 2-[1-(4-bromophenyl)-1-phenylethoxy]-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-[1-(4-bromophenyl)-1-phenylethoxy]-N,N-dimethyl-, hydrochloride (9CI), Ethylamine, 2-[(p-bromo-α-methyl-α-phenylbenzyl)oxy]-N,N-dimethyl-, hydrochloride (6CI,7CI,8CI), Bromadryl, Embramine hydrochloride, Mebrophenhydramine, Mebrophenhydramine hydrochloride, Mebryl
InChI formel InChI=1S/C18H22BrNO.ClH/c1-18(21-14-13-20(2)3,15-7-5-4-6-8-15)16-9-11-17(19)12-10-16;/h4-12H,13-14H2,1-3H3;1H
Løsningstype Neat
SMIL Cl.CN(C)CCOC(C)(c1ccccc1)c2ccc(Br)cc2
IUPAC navn 2-[1-(4-bromophenyl)-1-phenylethoxy]-N,N-dimethylethanamine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 384.74
Mængde 5 mg
Formel vægt 383.0652 g/mol