missing translation for 'onlineSavingsMsg'
Få mere at vide

Embelin, MedChemExpress

Artikelnummer. 16487358
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
Pakningsstørrelse:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16487358 10 mg -
16423387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16487358 Leverandør Medchem Express Leverandørnr. HY17473

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Embelin (Embelic acid), a potent, nonpeptidic XIAP inhibitor (IC50=4.1 μM), inhibits cell growth, induces apoptosis, and activates caspase-9 in prostate cancer cells with high levels of XIAP. Embelin blocks NF-kappaB signaling pathway leading to suppression of NF-kappaB-regulated antiapoptotic and metastatic gene products. Embelin also induces autophagic and apoptotic cell death in human oral squamous cell carcinoma cells.

Embelin (Embelic acid), a potent, nonpeptidic XIAP inhibitor (IC50=4.1 μM), inhibits cell growth, induces apoptosis, and activates caspase-9 in prostate cancer cells with high levels of XIAP. Embelin blocks NF-kappaB signaling pathway leading to suppression of NF-kappaB-regulated antiapoptotic and metastatic gene products. Embelin also induces autophagic and apoptotic cell death in human oral squamous cell carcinoma cells.

Tekniske data

Kemisk navn eller materiale Embelin
Fysisk form Solid
CAS 550-24-3
Farve Orange
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C17H26O4
Til brug med (applikation) Cancer-programmed cell death
Synonym Embelic acid Emberine NSC 91874
Opløselighedsinformation DMSO : ≥ 50 mg/mL (169.84 mM)
SMIL O=C1C(O)=C(CCCCCCCCCCC)C(C(O)=C1)=O
Molekylvægt (g/mol) 294.39
Formel vægt 294.39
Procent renhed 98.19%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.