missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Macherey-Nagel™ Elution Plate
Klik for at se tilgængelige muligheder
Samlet volumen (metrisk) Well:
370μ L
Pakningsstørrelse:
Pakke med 20
Beskrivelse
NucleoMag 96 Tissue is designed for the rapid parallel purification of genomic DNA from animal tissues, cell cultures and bacteria in a 96 well format. NucleoMag 96 Trace is designed for the purification of DNA from forensic samples (e.g. buccal swabs, dried blood spots, cigarette filters). NucleoMag 96 Plant is designed for the purification of genomic DNA from plant tissues in a 96 well format.
The magnetic bead technology provides a closed system therefore, the risk for cross contamination is greatly reduced.The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg NucleoMag SEP) or magnetic separators integrated into robotic workstations.Kit components: NucleoMag 96 Tissue – NucleoMag B-beads, buffers, Proteinase K; NucleoMag 96 Trace – NucleoMag B-beads, buffers, Proteinase K; NucleoMag 96 Plant – NucleoMag C-beads, buffers.
Rapid, economic purification of genomic DNA using highly reactive superparamagnetic beads, easily adapted to automated use
The magnetic bead technology provides a closed system therefore, the risk for cross contamination is greatly reduced.The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg NucleoMag SEP) or magnetic separators integrated into robotic workstations.Kit components: NucleoMag 96 Tissue – NucleoMag B-beads, buffers, Proteinase K; NucleoMag 96 Trace – NucleoMag B-beads, buffers, Proteinase K; NucleoMag 96 Plant – NucleoMag C-beads, buffers.
Rapid, economic purification of genomic DNA using highly reactive superparamagnetic beads, easily adapted to automated use
- Sample size: NucleoMag 96 Tissue – up to 20mg animal tissue, 1 x 107 cells or bacteria NucleoMag 96 Trace – buccal swabs, dried blood spots, cigarette filters NucleoMag 96 Plant – 20mg to 50mg plant tissue
- Typical yield: NucleoMag 96 Tissue/Plant – 10μg to 20μg DNA NucleoMag 96 Trace – 1μg to 3μg DNA
- A260/280 ≥1.6 to 1.9
- Concentration of sample possible
- Complete removal of PCR* inhibitors
- Small elution volumes (≥50μL)
- Yield does not depend on elution volume
- DNA ready to use in downstream applications
- Binding capacity: 0.4μg DNA per μL of NucleoMag bead suspension
- Size of beads: 1μm to 3μm
- Reduced carry-over of beads
Tekniske data
Tekniske data
| Format | Standard |
| Array | 8 x 12 |
| Antal Wells | 96 |
| Samlet volumen (metrisk) Well | 370μ L |
| Well Shape | Flad bund (F-brønd) |
| nr. pr. pakke | 20 |
| Mængde | 20/Pk. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion