Få mere at vide
Eltrombopag, TRC
Artikelnummer. 30134158
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134158 5 mg
17994625 100 mg
17991215 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134158 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5080005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 496775-61-2
Molekylær formel C25 H22 N4 O4
Molekylvægt (g/mol) 442.47
Synonym [1,1'-Biphenyl]-3-carboxylic acid, 3'-[2-[(2Z)-1-(3,4-dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-hydroxy-, [1,1'-Biphenyl]-3-carboxylic acid, 3'-[(2Z)-[1-(3,4-dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazino]-2'-hydroxy- (9CI), 3'-[2-[(2Z)-1-(3,4-Dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-hydroxy[1,1'-biphenyl]-3-carboxylic acid, Eltrombopag, SB 497115, Eltrombopag free acid, Eltrombopag free acid
IUPAC navn 3-[3-[(2Z)-2-[1-(3,4-dimethylphenyl)-3-methyl-5-oxopyrazol-4-ylidene]hydrazinyl]-2-hydroxyphenyl]benzoic acid
SMIL CC1=NN(C(=O)/C/1=N\Nc2cccc(c2O)c3cccc(c3)C(=O)O)c4ccc(C)c(C)c4
Tekniske data
Kemisk navn eller materiale Eltrombopag
Mængde 5 mg
InChI formel InChI=1S/C25H22N4O4/c1-14-10-11-19(12-15(14)2)29-24(31)22(16(3)28-29)27-26-21-9-5-8-20(23(21)30)17-6-4-7-18(13-17)25(32)33/h4-13,26,30H,1-3H3,(H,32,33)/b27-22-
Formel vægt 442.1641