Få mere at vide
Eleutheroside E, TRC
Artikelnummer. 30137332
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137332 10 mg
30134912 25 mg
30011010 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137332 Leverandør Toronto Research Chemicals Leverandørnr. TRCE50502510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 39432-56-9
Molekylær formel C34 H46 O18
Molekylvægt (g/mol) 742.72
IUPAC navn (2S,3R,4S,5S,6R)-2-[4-[(3R,3aS,6S,6aR)-6-[3,5-dimethoxy-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-3-yl]-2,6-dimethoxyphenoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL COc1cc(cc(OC)c1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]3OC[C@H]4[C@H]3CO[C@@H]4c5cc(OC)c(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c(OC)c5
Tekniske data
Kemisk navn eller materiale Eleutheroside E
Renhedsgrad noter HPLC
Mængde 10 mg
Synonym β-D-Glucopyranoside, [(1R,3aR,4S,6aS)-tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2,6-dimethoxy-4,1-phenylene) bis-, β-D-Glucopyranoside, (tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl)bis(2,6-dimethoxy-4,1-phenylene) bis-, [1R-(1α,3aβ,4β,6aα)]-, 1H,3H-Furo[3,4-c]furan, β-D-glucopyranoside deriv., Eleutheroside E, [(1R,3aR,4S,6aS)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2,6-dimethoxy-4,1-phenylene) bis-β-D-glucopyranoside
InChI formel InChI=1S/C34H46O18/c1-43-17-5-13(6-18(44-2)31(17)51-33-27(41)25(39)23(37)21(9-35)49-33)29-15-11-48-30(16(15)12-47-29)14-7-19(45-3)32(20(8-14)46-4)52-34-28(42)26(40)24(38)22(10-36)50-34/h5-8,15-16,21-30,33-42H,9-12H2,1-4H3/t15-,16+,21-,22-,23-,24-,25+,26+,27-,28-,29+,30-,33+,34+/m1/s1
Formel vægt 742.2684
Procent renhed >95%