Få mere at vide
La-Pha-Pack GmbH Electronic Handheld Crimper for 13 mm Flip-Off Caps
Artikelnummer. 16326218 Shop alle Thermo Scientific produkter
Skift visning
Til brug med (udstyr):
Crimp Caps (13 mm)
Artikelnummer. Til brug med (udstyr)
16326218 Crimp Caps (13 mm)
1 options
Artikelnummer. 16326218 Leverandør La-Pha-Pack GmbH Leverandørnr. 60180ECR13FO
Forventet forsendelse: 13-11-2026
Crimper includes a fully rechargeable lithium ion battery, universal power supply/recharger, and international plug adapters.
Crimper includes a fully rechargeable lithium ion battery, universal power supply/recharger, and international plug adapters.
  • Electronic Handheld Crimper
  • For use with 13 mm Crimp Caps
  • Fully rechargeable lithium ion battery
  • Universal power supply/recharger
  • International plug adapters
TRUSTED_SUSTAINABILITY
Tekniske data
Produkttype Crimper Jaw
Til brug med (udstyr) Crimp Caps (13 mm)
Batteritype Lithium-ion
Diameter (metrisk) 13 mm
Inkluderer Power Supply/Recharger, and International Plug Adaptors