missing translation for 'onlineSavingsMsg'
Learn More

Elaidic Acid Methyl Ester, TRC

Product Code. 30373527
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30373527 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373527 Supplier Toronto Research Chemicals Supplier No. TRCE500205100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Elaidic acid methyl ester
CAS 1937-62-8
Molekylær formel C19 H36 O2
Mængde 100 mg
Synonym 9-Octadecenoic acid, methyl ester, (E)-, Elaidic acid, methyl ester (6CI,8CI), 9(E)-Octadecenoic acid methyl ester, Methyl 9(E)-octadecenoate, Methyl elaidate, Methyl trans-9-octadecenoate, trans-Oleic acid methyl ester, Elaidic Acid Methyl Ester, 9-Octadecenoic acid, methyl ester, (9E)-
InChI formel InChI=1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+
SMIL CCCCCCCC\C=C\CCCCCCCC(=O)OC
IUPAC navn methyl (E)-octadec-9-enoate
Molekylvægt (g/mol) 296.49
Formel vægt 296.2715
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.