Få mere at vide
Elagolix Methoxy-d3 Sodium Salt, TRC
Artikelnummer. 30370534
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30370534 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370534 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5010241MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Elagolix Methoxy-d3 Sodium Salt
CAS 2233595-41-8
Molekylær formel C32H26D3F5N3NaO5
Mængde 1 mg
Synonym 4-[[(1R)-2-[5-[2-Fluoro-3-(methoxy-d3)phenyl]-3-[[2-fluoro-6-(trifluoromethyl)phenyl]methyl]-3,6-dihydro-4-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-1-phenylethyl]amino]-butanoic Acid Sodium Salt
InChI formel InChI=1S/C32H30F5N3O5.Na/c1-19-28(21-11-6-14-26(45-2)29(21)34)30(43)40(18-25(20-9-4-3-5-10-20)38-16-8-15-27(41)42)31(44)39(19)17-22-23(32(35,36)37)12-7-13-24(22)33;/h3-7,9-14,25,38H,8,15-18H2,1-2H3,(H,41,42);/q;+1/p-1/t25-;/m0./s1/i2D3;
SMIL O=C(N1C[C@H](NCCCC(O[Na])=O)C2=CC=CC=C2)C(C3=CC=CC(OC([2H])([2H])[2H])=C3F)=C(C)N(CC4=C(C(F)(F)F)C=CC=C4F)C1=O
Molekylvægt (g/mol) 656.59