Learn More
Chemsplash Eka Jacket A/S with Collar Zip and Adhesive Flap
GREENER_CHOICE
Disposable Zip Jacket are made of soft and microporous Chemsplash fabric. Strong and resistant to chemical splash and dust.
1290.00 DKK
Specifications
Product Type Short Lab Coat
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Zipper
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30058475
GREENER_CHOICE
View Documents
Chemsplash
2606.400.004
Small
1290.00 DKK
Pack of 25
Only null left
Estimated Shipment: 16-03-2026
Add to basket
30058216
GREENER_CHOICE
View Documents
Chemsplash
2606.400.005
Medium
1290.00 DKK
Pack of 25
Only null left
Estimated Shipment: 16-03-2026
Add to basket
30058245
GREENER_CHOICE
View Documents
Chemsplash
2606.400.006
Large
1290.00 DKK
Pack of 25
Only null left
Estimated Shipment: 16-03-2026
Add to basket
30058463
GREENER_CHOICE
View Documents
Chemsplash
2606.400.007
X-Large
1290.00 DKK
Pack of 25
Only null left
Estimated Shipment: 16-03-2026
Add to basket
30058422
GREENER_CHOICE
View Documents
Chemsplash
2606.400.008
2X-Large
1290.00 DKK
Pack of 25
Only null left
Estimated Shipment: 16-03-2026
Add to basket
30058215
GREENER_CHOICE
View Documents
Chemsplash
2606.400.009
3X-Large
1290.00 DKK
Pack of 25
Only null left
Estimated Shipment: 16-03-2026
Add to basket
Description
Description
Chemsplash Disposable Zip Jacket
  • Chemsplash 55GSM Type PB 6B Fabric
  • Long Sleeves and Collar
  • Covered Zip
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting Fabric
  • PFAS Free
  • Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9
Description: Chemsplash Disposable Zip Jacket is made of soft and microporous Chemsplash fabric. It is strong and resistant to chemical splash and dust. Lint-free and anti-static in accordance with EN1149-5:2008. Chemsplash accessories are used to protect the wearer from the environment or the environment from the wearer. The range is designed to allow for maximum comfort and wearability. Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9.
Specifications
Specifications
Short Lab Coat
Microporous Composite Fabric
Zipper
Single-use
Liquids
Yes
Unisex
White
Elastic
Non-sterile
None
Class 100-100,000 (ISO 5-8)
Documents
Product Certifications
Product Content Correction
Product Title
Chemsplash Eka Jacket A/S with Collar Zip and Adhesive Flap