missing translation for 'onlineSavingsMsg'
Learn More

Chemsplash Eka 55 Disposable Coverall Type 5/6
GREENER_CHOICE

Eka 55 Disposable Coverall is made with 55 gsm microporous laminated non-linting fabric.

1225.00 DKK

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30058145
GREENER_CHOICE
View Documents
Chemsplash
2756.400.012
6X-Large
1225.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058136
GREENER_CHOICE
View Documents
Chemsplash
2757.400.011
5X-Large
1225.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058148
GREENER_CHOICE
View Documents
Chemsplash
2758.400.010
4X-Large
1225.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
Description

Description

Chemsplash Eka 55 Disposable Coverall

  • 55GSM Microporous Laminate Fabric
  • Elasticated 3pce Hood and Back
  • Elasticated Cuffs
  • Two Way Zip
  • Adhesive Zip Flap
  • Elasticated Ankles
  • Silicone and Latex Free
  • Non-Linting Fabric
  • Anti-Static
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9

Description: The Chemsplash Eka 55 Disposable Coverall is made from 55GSM microporous laminated non-linting fabric. It provides excellent value for laminated Cat III Type 5B/6B protection, particularly suited for applications where the coverall is not used for prolonged periods but still needs to offer high protection against liquid chemical splashes. Featuring an elasticated back, hood, cuffs, and ankles, along with an adhesive zip flap for additional sealing, this suit ensures both comfort and protection. The Chemsplash Eco fabric is Anti-Static to EN1149-5:2008 and non-linting, making it ideal for environments where garment fibres must not contaminate the operating space.

Specifications

Specifications

Cleanroom Coveralls
Microporous Composite Fabric
Single-use
Elastic
Hooded
Ties
Non-sterile
Yes
Class 100-100,000 (ISO 5-8)
Unisex
White
Elastic
Elastic
3-panel
Stitched
Liquid
None
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.