missing translation for 'onlineSavingsMsg'
Learn More

Chemsplash Eka 55 C/All, Type 5/6, A/S Laminate, Hood Orange
GREENER_CHOICE

Eka55 Coverall 55 gsm microporous laminated fabric. Has an elasticated back, hood, cuffs and ankles, and an adhesive cover flap for additional sealing.

1180.00 DKK

Specifications

Product Type Cleanroom Coveralls
Material Microporous Composite Fabric
Color Orange
Disposable Single-use
Waist Style Elastic
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30058144
GREENER_CHOICE
View Documents
Chemsplash
2636.450.004
Small
1180.00 DKK
Pack of 25
Only null left
Estimated Shipment: 31-07-2025
Add to basket
 
30058167
GREENER_CHOICE
View Documents
Chemsplash
2636.450.005
Medium
1180.00 DKK
Pack of 25
Only null left
Estimated Shipment: 31-07-2025
Add to basket
 
30058328
GREENER_CHOICE
View Documents
Chemsplash
2636.450.006
Large
1180.00 DKK
Pack of 25
Only null left
Estimated Shipment: 31-07-2025
Add to basket
 
30058282
GREENER_CHOICE
View Documents
Chemsplash
2636.450.007
X-Large
1180.00 DKK
Pack of 25
Only null left
Estimated Shipment: 31-07-2025
Add to basket
 
30058392
GREENER_CHOICE
View Documents
Chemsplash
2636.450.008
2X-Large
1180.00 DKK
Pack of 25
Only null left
Estimated Shipment: 31-07-2025
Add to basket
 
30058160
GREENER_CHOICE
View Documents
Chemsplash
2636.450.009
3X-Large
1180.00 DKK
Pack of 25
Only null left
Estimated Shipment: 31-07-2025
Add to basket
 
Description

Description

Chemsplash Eka55 Coverall

  • 55GSM Microporous Laminate Fabric
  • Elasticated Hood, Back and Cuffs
  • Elasticated Ankles
  • Two Way Zip With Adhesive Zip Flap
  • Anti-Static
  • Non Linting
  • Silicone, Latex Free and PFAS Free
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9

Description: The Chemsplash Eka55 Coverall is made from 55gsm microporous laminated fabric. It offers users a better value laminated Cat III Type 5/6 coverall for applications where the suit isn't used for prolonged periods but still requires high protection from liquid chemical splashes. The suit features an elasticated back, hood, cuffs, and ankles, as well as an adhesive cover flap for additional sealing. The Chemsplash Eco fabric is anti-static to EN1149-5:2008 and non-linting, making it ideal for environments where contamination from garment fibres must be minimized.

Specifications

Specifications

Cleanroom Coveralls
Orange
Elastic
Hooded
Ties
Non-sterile
Yes
Class 100-100,000 (ISO 5-8)
Microporous Composite Fabric
Single-use
Elastic
3-panel
Stitched
Liquid
None
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.