missing translation for 'onlineSavingsMsg'
Learn More

Chemsplash Eka 55 C/All with Feet, Type 5/6, A/S Laminate, Hood
GREENER_CHOICE

Eka55 Coverall with feet, is made from 55 gsm microporous, laminated, non-linting fabric.

1255.00 DKK

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30058155
GREENER_CHOICE
View Documents
Chemsplash
2626.400.004
Small
1255.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058228
GREENER_CHOICE
View Documents
Chemsplash
2626.400.005
Medium
1255.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058249
GREENER_CHOICE
View Documents
Chemsplash
2626.400.006
Large
1255.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058347
GREENER_CHOICE
View Documents
Chemsplash
2626.400.007
X-Large
1255.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058363
GREENER_CHOICE
View Documents
Chemsplash
2626.400.008
2X-Large
1255.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058223
GREENER_CHOICE
View Documents
Chemsplash
2626.400.009
3X-Large
1255.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
Description

Description

Chemsplash Eka55 Coverall

  • 55GSM Microporous Laminate Fabric
  • With Feet Attached
  • Elasticated 3pce Hood, Back and Cuffs
  • Elasticated Ankles
  • Two Way Zip With Adhesive Zip Flap
  • Anti-Static
  • Non Linting Fabric
  • Silicone, Latex Free and PFAS Free
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9

Description: Chemsplash Eka55 Coverall with feet is made from 55gsm microporous laminated non-linting fabric. It offers users a better value laminated Cat III Type 5B/6B coverall for those applications where the suit isn't used for prolonged periods but where a high level of protection to liquid chemical splashes is still required. This suit has an elasticated back, hood, cuffs and ankles, and an adhesive cover flap for additional sealing. Chemsplash Eco fabric is Anti-Static to EN1149-5:2008 and non-linting, therefore ideal for use in wide-ranging applications where the operating environment must not be contaminated with garment fibres.

Specifications

Specifications

Cleanroom Coveralls
Microporous Composite Fabric
Single-use
Attached Boots
Hooded
Ties
Non-sterile
Yes
Class 100-100,000 (ISO 5-8)
Unisex
White
Elastic
Elastic
3-panel
Stitched
Liquid
None
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.