Få mere at vide
Eicosyl(trimethyl)azanium Bromide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Eicosyl(trimethyl)azanium Bromide |
| CAS | 7342-61-2 |
| Molekylær formel | C23 H50 N . Br |
| Mængde | 100 mg |
| Synonym | N,N,N-Trimethyl-1-Eicosanaminium Bromide, Eicosyltrimethyl-ammonium Bromide, Eicosyltrimethylammonium bromide, Eicosanyltrimethylammonium Bromide |
| InChI formel | InChI=1S/C23H50N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(2,3)4;/h5-23H2,1-4H3;1H/q+1;/p-1 |
| SMIL | [Br-].CCCCCCCCCCCCCCCCCCCC[N+](C)(C)C |
| IUPAC navn | icosyl(trimethyl)azanium;bromide |
| Molekylvægt (g/mol) | 420.554 |
| Formel vægt | 419.313 |