Få mere at vide
Eicosanoic-20,20,20-d3 Acid, CDN
Artikelnummer. 30320208
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320208 0.1 g 0.1g
30308813 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320208 Leverandør CDN Leverandørnr. CDND52540.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Eicosanoic Acid-20,20,20-D3
CAS 202480-70-4
Molekylær formel C20 D3 H37 O2
Mængde 0.1 g
Synonym Eicosanoic-20,20,20-d3 acid (9 CI), Eicosanoic Acid-20,20,20-D3, Eicosanoic acid-d3, Arachidic-20,20,20-d3 acid, Arachidic acid-20,20,20-d3, Arachidic acid-ω-d3
InChI formel InChI=1 S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-19H2,1H3,(H,21,22)/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCCCCC(=O)O
IUPAC navn 20,20,20-trideuterioicosanoic acid
Molekylvægt (g/mol) 315.5489
Formel vægt 315.3217 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity