missing translation for 'onlineSavingsMsg'
Learn More

EGFR Recombinant Human Protein, His Tag, Active

EGFR Recombinant Human Protein (His Tag, Active)

Brand:  Invitrogen 10001H08H250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15294475

  • 8010.00 DKK / 250µg


Estimated Shipment
22-09-2020
Add to basket

Description

Description

Epidermal growth factor receptor (EGFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant protein

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 645) of human EGFR (NP_005219) fused to a polyhistidine tag at the C-terminus.

Activity: Measure by its ability to bind with human EGF in a functional ELISA.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

Specifications

Specifications

Mammalian
Growth factor receptors, receptor tyrosine kinases
Protein assays and analysis
Lyophilized
EGFR Recombinant Human Protein
5 x 50μg
For research use only. Not for use in diagnostic procedures
EGFR, ErbB, HER1, mENA, ErbB1, PIG61
5 Vials of lyophilized protein
EGF receptor (ErbB)
Human
Growth factors and receptors, kinases and inhibitors
Assay standard
Lyophilized
69.8kDa
>95% by SDS-PAGE
–20°C
5 Pack(s)
EGFR Recombinant Human Protein
EGFR
EGFR Recombinant Human Protein
His Tag
Videos
Documents