Learn More
Invitrogen™ EGFR (ErbB1) [T790M] Recombinant Human Protein
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors.
Brand: Invitrogen™ PV4804
Additional Details : Weight : 0.01000kg
Description
Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The T790M mutant of EGFR has arisen in the clinics and displays resistance to Erlotinib (Tarceva) and Gefitinib (Iressa).
Specifications
NP_005219.2; T790M | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
EGFR (ErbB1) T790M | |
Human | |
GST Tag | |
Frozen |
Nuclear Receptor Assay | |
90.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Baculovirus |