missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EGFR [d746-750] Recombinant Human Protein

Human EGF-R, C-terminal fragment, amino acids H672-A1210 (as in GenBank entry NM_005228.3), amino acids 746-750 deleted, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.

Brand:  Invitrogen™ PV6178

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15478843

  • 3660.00 DKK / 10µg
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
88.6kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
aa 672-1210
Recombinant
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol
10μg
ERBB, ERBB1
Human
GST Tag, His Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers