Få mere at vide
Efavirenz USP Related Compound A, Mikromol™
Artikelnummer. 30050530
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050530 25 mg
30050400 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050530 Leverandør Mikromol Leverandørnr. MM0919.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Efavirenz USP Related Compound A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol
CAS 209414-27-7
Analyt- eller komponentnavne (S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C13 H11 Cl F3 N O
Certificeringer/compliance ISO 17025
Synonym (alphaS)-2-Amino-5-chloro-alpha-(cyclopropylethynyl)-alpha-(trifluoromethyl)benzenemethanol, (S)-1-(2-Amino-5-chlorophenyl)-1-(trifluoromethyl)-3-cyclopropyl-2-propyn-1-ol, (S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluoro-3-butyn-2-ol, (alphaS)-2-Amino-5-chloro-alpha-(cyclopropylethynyl)-alpha-(trifluoromethyl)benzenemethanol, Efavirenz Related Compound A (USP), Efavirenz Related Compound A
InChI formel InChI=1S/C13H11ClF3NO/c14-9-3-4-11(18)10(7-9)12(19,13(15,16)17)6-5-8-1-2-8/h3-4,7-8,19H,1-2,18H2/t12-/m0/s1
SMIL Nc1ccc(Cl)cc1[C@@](O)(C#CC2CC2)C(F)(F)F
IUPAC navn (2S)-2-(2-amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol
Molekylvægt (g/mol) 289.68
Mængde 25 mg
Formel vægt 289.0481 g/mol