missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium Salt Solution, 0.01M, Chem-Lab

Catalog Number p-8014430
Click to view available options
Quantity:
1 L
5 L
This item is not returnable. View return policy
6381-92-6
C10H18N2Na2O10
372.24
MFCD00150037,MFCD00003541
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
This item is not returnable. View return policy
6381-92-6
C10H18N2Na2O10
372.24
MFCD00150037,MFCD00003541
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Used in electrophoresis and molecular biology applications. EDTA Disodium Salt Solution, 0.01M, Chem-Lab is a chelator of divalent cations for use in TH titrations. Solution contains 3.723g EDTA diNa • 2H2O per liter of water (0.01 M).

EDTA Disodium Salt Dihydrate is a chelator of divalent cations. It inhibits enzymes, such as metalloproteases, that require divalent cations for activity. It is also used in many enzyme buffers and as an enzyme inactivator at high concentrations.
Melting Point 0°C
Color Colorless
Density 1g/cm3
pH 4 to 5
Boiling Point 100°C
Odor Odorless
Quantity 1 L
Solubility Information Soluble
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Formula Weight 372.24g/mol
Concentration or Composition (by Analyte or Components) 0.01mol/L (± 0.00004, 20°C)
Physical Form Liquid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'