Få mere at vide
Epredia™ Edge-Rite™ Disposable Microtome Blades
Shop alle Epredia produkter
Profil:
Høj
Lav
Pakningsstørrelse:
Pakke med 50
Beskrivelse
- Surgical-grade steel
- High- and low-profile blades are specially coated to increase longevity and optimize section quality
Low profile blades:
- Blades are 20% thicker than traditional blades to increase knife edge stability, which allows for excellent ribboning and facilitates usage in cryostats
- Blades are 5% longer, allowing for more sections per blade
- Blades fit any low-profile blade holder
Tekniske data
Tekniske data
| Produkttype | Disposable Microtome Blade |
| Profil | Lav |
| Materiale | Surgical-grade Steel |
| Engangs | Single-use |