Learn More
Epredia™ Edge-Rite™ Disposable Microtome Blades
Experience consistent quality from blade to blade and sample to sample with Epredia™ Edge-Rite™ disposable microtome blades.
Brand: Epredia™ 4275H
1015.93 DKK valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Surgical-grade steel
- High- and low-profile blades are specially coated to increase longevity and optimize section quality
Low profile blades:
- Blades are 20% thicker than traditional blades to increase knife edge stability, which allows for excellent ribboning and facilitates usage in cryostats
- Blades are 5% longer, allowing for more sections per blade
- Blades fit any low-profile blade holder
Specifications
Disposable Microtome Blade | |
High | |
Surgical-grade Steel | |
Yes |