Beskrivelse
- High durability
- Nitrile foam coating delivers abrasion resistance and grip
- Breathable liner allows high level of comfort for end users
- Secure grip in dry and slightly oily environments
- Made of a seamless liner,
- Alternative to heavy duty leather and polyurethane palm dipped cut resistant knitted gloves
- Automotive, construction, glass manufacturing, machinery, equipment, metal fabrication
Tekniske data
Tekniske data
| ANSI Abrasion Rating | ANSI 5 |
| Certificeringer/overholdelse (Europa) | CE, EN 420:2003, A1:2009, Cat II |
| Konstruktion | Knit |
| Belægningsmateriale | Nitrile Foam |
| Farve | Gray, Black |
| Bedømmelse af skæremodstand | ANSI A3 |
| Produkttype | Cut-resistant Gloves |
| Størrelse | 7 |
| EN388 Standard | EN 388:4X43C |
| Til brug med (applikation) | General maintenance, hardware assembly, machining, stamping, cutting, molding, plastic injections, raw material handling, seat and frame build |
| Vis mere |