Få mere at vide
EDDHSA, TRC
Artikelnummer. 30133609
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133609 100 mg
30009437 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133609 Leverandør Toronto Research Chemicals Leverandørnr. TRCE335105100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57368-07-7
Molekylær formel C18H20N2O12S2
Molekylvægt (g/mol) 520.49
Synonym α,α'-(1,2-Ethanediyldiimino)bis[2-hydroxy-5-sulfo-benzeneacetic Acid, N,N-Ethylenebis[2-(2-hydroxy-5-sulfophenyl)glycine, N,N'-Ethylenebis[2-(2-hydroxy-5-sulfophenyl)glycine
SMIL Fc1nc2c(ncn2)c(N)n1
Tekniske data
Kemisk navn eller materiale EDDHSA
Mængde 100 mg
InChI formel InChI=1S/C18H20N2O12S2/c21-13-3-1-9(33(27,28)29)7-11(13)15(17(23)24)19-5-6-20-16(18(25)26)12-8-10(34(30,31)32)2-4-14(12)22/h1-4,7-8,15-16,19-22H,5-6H2,(H,23,24)(H,25,26)(H,27,28,29)(H,30,31,32)