Få mere at vide
Edaravone D5, TRC
Artikelnummer. 30371396
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30371396 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371396 Leverandør Toronto Research Chemicals Leverandørnr. TRCE1228725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Edaravone D5
CAS 1228765-67-0
Molekylær formel C10 D5 H5 N2 O
Mængde 5 mg
Synonym 2,4-Dihydro-5-methyl-2-(phenyl-2,3,4,5,6-d5)-3H-pyrazol-3-one
InChI formel InChI=1S/C10H10N2O/c1-8-7-10(13)12(11-8)9-5-3-2-4-6-9/h2-6H,7H2,1H3/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(N2N=C(C)CC2=O)c([2H])c1[2H]
IUPAC navn 5-methyl-2-(2,3,4,5,6-pentadeuteriophenyl)-4H-pyrazol-3-one
Molekylvægt (g/mol) 179.23
Formel vægt 179.111