missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
Velp Scientifica ECODryBlock is a modern, versatile dry block heater designed for precise temperature control and flexible laboratory workflows.
Key Features
- Modern, intuitive interface with a 7 in. ControlPad featuring a capacitive touchscreen and durable, chemical-resistant glass surface.
- Flexible configuration: one ControlPad can manage up to two Heating Modules, each accommodating up to two aluminum blocks (95 x 76 x 51 mm).
- Independent control of temperature (up to 165°C) and timers for each Module.
- Wide selection of interchangeable aluminum blocks suitable for various vials, tubes, and cuvettes.
- Optional Transparent lid creates a warm air jacket, minimizing heat loss and ensuring uniform temperature distribution.
- External Pt100 and Pt1000 probes allow precise temperature monitoring and control directly in the block or sample vessel.
Applications
- Chemical Oxygen Demand (COD) in water samples.
- DNA/RNA extraction and purification
- Enzymatic reactions
- Activation of cell cultures
- DNA and protein denaturation
- Assay preparation and clinical tests
- Melting and boiling point determination
- Culture media tests
- Other analytical protocols
Accessory Included
- Transparent lid for ECODryBlock
- COD-W24-D160-A Block
Tekniske data
Tekniske data
| Certificeringer/compliance | CE, UKCA |
| Controller type | ControlPad |
| Autoklaverbar | Not Autoclavable |
| Temperaturområde (metrisk) | 5°C to 165°C |
| Type | Dry Block Heater |
| Vise | Digital |
| Inkluderer | Transparent lid for ECODryBlock, COD-W24-D160-A Block |
| Temperatur nøjagtighed | 0.5°C |
| Temperaturstabilitet | ±0.5°C at 165°C |
| Temperaturensartethed | ±0.5°C at 165°C |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?