Learn More
LevGo™ Eco-smartFunnel™ Disposable Paper Lab Funnel, Fsc Mix Credit, Micro
Product Code. 30696095
Change view
Click to view available options
Material:
Paper
Unit Size:
Pack of 25
Product Code. Material Unit Size
30696095 Paper Pack of 25
1 options
Product Code. 30696095 Supplier LevGo™ Supplier No. 19620
  • ONLINE EXCLUSIVE PRICE
    213.00 DKK
    200.07 DKK / Pakke med 25
    Spare 12.93 DKK
    6% Slukket
Forventet forsendelse: 07-12-2026
Disposable funnel that provides a clean transfer every time.
Eco Friendly Disposable Paper Laboratory Funnel
  • Use with virtually any liquid or powder (not recommended for hot liquids)
  • Often used to collect sputum samples
  • Environmentally friendly paper product that will naturally biodegrade in the standard municipal waste stream
  • Made of FSC™ Certified, FDA food grade, clay coated heavy grade paper
  • Innovative backrest allows hands - free use when pouring viscous liquids
  • Maximum flexibility - cut stem to any desired opening size or shape
  • Apply a minimum of pressure and the funnel will stay in position
  • Made in the USA
Specifications
Material Paper
Product Type Addition Funnel
Material (Membrane) Paper