missing translation for 'onlineSavingsMsg'
Learn More
BD Eclipse™ Hypodermic Injection Needle with SmartSlip™ Technology Product Code.: 15410875

BD Eclipse™ Hypodermic Injection Needle with SmartSlip™ Technology

Product Code. 15410875
18 G, Pack of 100
Click to view available options
Needle Gauge:
18 G
21 G
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15410875

Brand: BD 302437

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensures protection from needlestick injuries after injection. The BD Eclipse™ Hypodermic Injection Needle features the same safety mechanism as the proven BD Eclipse™ Blood Collection Needle, with SmartSlip™ Technology to ensure a secure connection to Luer slip syringes.

  • Unique SmartSlip technology helps to ensure a secure connection to Luer slip syringes
  • Bevel oriented to safety cover accommodates low angle injections
  • One technology for skin injection and blood collection
  • Requires minimal change in injection technique
  • The hub incorporates a plastic clip to encourage user to attach needle more securely to Luer slip syringe
  • Wide textured finger pad provides safe activation area for thumb or forefinger
  • Integral safety cover and extended sidewall to enhance protection
  • Needle remains locked inside activated needle cover
  • Activates with a touch of a finger – no hard surface required for activation
  • Colour coded hub for easy needle size identification
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EU directive
Color Pink
Needle Gauge 18 G
Diameter (Metric) Outer Needle 1.2 mm
For Use With (Application) Hypodermic injections
Includes Safety cover
Needle Point Style Bevel
For Use With (Equipment) Luer Slip Syringe
Connection Type Luer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Eclipse™ Hypodermic Injection Needle with SmartSlip™ Technology >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.