Learn More
Ebro Electronic™ Accessories for EBI 20 Data Logger Series
Data logger accessories to aid measurement of temperature and / or humidity during transportation and in warehouses
Brand:  Ebro Electronic™ 1601-0020
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10028852
  • 849.54 DKK / Each
Estimated Shipment: 26-08-2024
to see stock.
Add to basket
Description
Description
  • Connected to the PC and supplied with power from there by means of a USB cable
  • Supplied with Interface, USB cable, software Winlog.basic, instruction manual
  • Clean with a slightly damp cloth only
Order Info
Includes
Specifications
Specifications
Interface
Not Calibrated
Interface EBI 20-IF, USB-cable, Software Winlog.basic, instructions for use.
EBI 20-IF
No Display
None
EBI 20-IF
External