Få mere at vide
Tocris Bioscience™ EB 47
GREENER_CHOICE
Artikelnummer. 18054362
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18054362 10 mg
18034542 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18054362 Leverandør Tocris Bioscience™ Leverandørnr. 4140/10
Potent PARP-1 inhibitor
Potent inhibitor of PARP-1 (IC50 = 45 nM). Reduces infarct volume in both a rat transient middle cerebral arterial occlusion model and a cardiac reperfusion model.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1190332-25-2
Molekylær formel C24H29Cl2N9O6
Tekniske data
Kemisk navn eller materiale EB 47
Mængde 10 mg
Formel vægt 610.45
Renhed >99%
Molekylvægt (g/mol) 610.453