Få mere at vide
STEMCELL Technologies EasySep™ RBC Depletion Reagent for RoboSep
Mængde:
100 mL Whole Blood
Beskrivelse
- The EasySep™ RBC Depletion Reagent is designed to immunomagnetically deplete red blood cells (RBCs) from fresh human peripheral blood, umbilical cord blood, bone marrow or leukapheresis samples by negative selection
- RBCs are targeted with Tetrameric Antibody Complexes recognizing Glycophorin A and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Unwanted cells remain in the tube while desired cells are poured off
- This product replaces the EasySep™ Human Glycophorin A Depletion Kit (Catalog #18352) for even faster cell isolations
Tekniske data
Tekniske data
Mængde | 100 mL Whole Blood |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
STEMCELL Technologies EasySep™ RBC Depletion Reagent for RoboSep
Ser du en mulighed for forbedring?Del en indholdskorrektion